Läs mer
N-Benzyl-L-serine, Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Benzyl-L-Serine, Methyl Ester |
| CAS | 123639-56-5 |
| Molekylformel | C11 H15 N O3 |
| Kvantitet | 2 g |
| Synonym | L-Serine, N-(phenylmethyl)-, methyl ester, (S)-Methyl 2-(benzylamino)-3-hydroxypropanoate, N-Benzyl-L-serine methyl ester, (S)-N-Benzylserine methyl ester, Methyl N-benzyl-L-serinate, Methyl (2S)-2-(benzylamino)-3-hydroxypropanoate |
| InChI formel | InChI=1S/C11H15NO3/c1-15-11(14)10(8-13)12-7-9-5-3-2-4-6-9/h2-6,10,12-13H,7-8H2,1H3/t10-/m0/s1 |
| LEDER | COC(=O)[C@H](CO)NCc1ccccc1 |
| IUPAC-namn | methyl (2S)-2-(benzylamino)-3-hydroxypropanoate |
| Molekylvikt (g/mol) | 209.24 |
| Formel vikt | 209.1052 |