Läs mer
N-Benzyl-L-tyrosine, TRC
Produktkod. 30342656
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30342656 100 mg
1 options
Produktkod. 30342656 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB296200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyl-L-tyrosine
CAS 75768-66-0
Molekylformel C16H17NO3
Kvantitet 100 mg
Synonym N-(Phenylmethyl)-L-tyrosine
InChI formel InChI=1S/C16H17NO3/c18-14-8-6-12(7-9-14)10-15(16(19)20)17-11-13-4-2-1-3-5-13/h1-9,15,17-18H,10-11H2,(H,19,20)/t15-/m0/s1
LEDER OC(=O)[C@H](Cc1ccc(O)cc1)NCc2ccccc2
IUPAC-namn (2S)-2-(benzylamino)-3-(4-hydroxyphenyl)propanoic acid
Molekylvikt (g/mol) 271.32
Formel vikt 271.1208