Läs mer
N-Benzyl N-Demethyl Trimebutine, TRC
Produktkod. 30339721
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30339721 1 mg
1 options
Produktkod. 30339721 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2345901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyl N-Demethyl Trimebutine
CAS 1329834-94-7
Molekylformel C28H33NO5
Kvantitet 1 mg
Synonym 3,4,5-Trimethoxybenzoic Acid 2-(N-Benzyl-methylamino)-2-phenylbutyl Ester, N-Benzyl N-Monodesmethyltrimebutine
InChI formel InChI=1S/C28H33NO5/c1-6-28(23-15-11-8-12-16-23,29(2)19-21-13-9-7-10-14-21)20-34-27(30)22-17-24(31-3)26(33-5)25(18-22)32-4/h7-18H,6,19-20H2,1-5H3
LEDER CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)Cc2ccccc2)c3ccccc3
IUPAC-namn [2-[benzyl(methyl)amino]-2-phenylbutyl] 3,4,5-trimethoxybenzoate
Molekylvikt (g/mol) 463.57
Formel vikt 463.2359