Läs mer
(N-Benzyl-N-ethyl) Amino-3-Hydroxyacetophenone Hydrochloride, TRC
Produktkod. 30332390
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30332390 1 g
1 options
Produktkod. 30332390 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6695601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzyletilefrone Hydrochloride)
CAS 55845-90-4
Molekylformel C17 H19 N O2 . Cl H
Kvantitet 1 g
Synonym 2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride, Etilefrine Hydrochloride Imp. D (EP) as Hydrochloride, Benzyletilefrone Hydrochloride
InChI formel InChI=1S/C17H19NO2.ClH/c1-2-18(12-14-7-4-3-5-8-14)13-17(20)15-9-6-10-16(19)11-15;/h3-11,19H,2,12-13H2,1H3;1H
LEDER Cl.CCN(CC(=O)c1cccc(O)c1)Cc2ccccc2
IUPAC-namn 2-[benzyl(ethyl)amino]-1-(3-hydroxyphenyl)ethanone;hydrochloride
Molekylvikt (g/mol) 305.8
Formel vikt 305.1183