missing translation for 'onlineSavingsMsg'
Learn More

N'-Benzyl-N,N-dimethylethylenediamine, 97%, Thermo Scientific™

Catalog Number p-7043855
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
103-55-9
C11H18N2
178.28
MFCD00008334
LLSJAFHDYCTFCM-UHFFFAOYSA-N
n'-benzyl-n,n-dimethylethylenediamine, 1,2-ethanediamine, n,n-dimethyl-n'-phenylmethyl, n,n-dimethyl-n'-benzylethylenediamine, benzyl 2-dimethylamino ethyl amine, n'-benzyl-n,n-dimethyl-ethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-n2-phenylmethyl, n'-benzyl-n,n-dimethylethane-1,2-diamine, n'-benzyl-n,n-dimethyl-ethylenediamine, n1-benzyl-n2,n2-dimethyl-1,2-ethanediamine, n1-benzyl-n2,n2-dimethylethane-1,2-diamine
66029
N-benzyl-N',N'-dimethylethane-1,2-diamine
CN(C)CCNCC1=CC=CC=C1
This item is not returnable. View return policy
103-55-9
C11H18N2
178.28
MFCD00008334
LLSJAFHDYCTFCM-UHFFFAOYSA-N
n'-benzyl-n,n-dimethylethylenediamine, 1,2-ethanediamine, n,n-dimethyl-n'-phenylmethyl, n,n-dimethyl-n'-benzylethylenediamine, benzyl 2-dimethylamino ethyl amine, n'-benzyl-n,n-dimethyl-ethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-n2-phenylmethyl, n'-benzyl-n,n-dimethylethane-1,2-diamine, n'-benzyl-n,n-dimethyl-ethylenediamine, n1-benzyl-n2,n2-dimethyl-1,2-ethanediamine, n1-benzyl-n2,n2-dimethylethane-1,2-diamine
66029
N-benzyl-N',N'-dimethylethane-1,2-diamine
CN(C)CCNCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.922
Boiling Point 255°C
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula C6H5CH2NHCH2CH2N(CH3)2
Refractive Index 1.509
Quantity 5 g
UN Number UN2735
Beilstein 2641339
Formula Weight 178.28
Percent Purity 97%
Chemical Name or Material N'-Benzyl-N,N-dimethylethylenediamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'