Läs mer
N-Benzyl Salbutamol Acetonide Methyl Ether, TRC
Produktkod. 30332741
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30332741 100 mg
1 options
Produktkod. 30332741 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB289025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyl Salbutamol Acetonide Methyl Ether
CAS 1797879-98-1
Molekylformel C24H33NO3
Kvantitet 100 mg
Synonym alpha-[[N-(1,1-Dimethylethyl)-N-benzylamino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol Methyl Ether
InChI formel InChI=1S/C24H33NO3/c1-23(2,3)25(15-18-10-8-7-9-11-18)16-22(26-6)19-12-13-21-20(14-19)17-27-24(4,5)28-21/h7-14,22H,15-17H2,1-6H3
LEDER COC(CN(Cc1ccccc1)C(C)(C)C)c2ccc3OC(C)(C)OCc3c2
IUPAC-namn N-benzyl-N-[2-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)-2-methoxyethyl]-2-methylpropan-2-amine
Molekylvikt (g/mol) 383.52
Formel vikt 383.246