Läs mer
N-Benzyldiacetamide, TRC
Produktkod. 30343672
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30343672 25 mg
1 options
Produktkod. 30343672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB28872525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyldiacetamide
CAS 3027-02-9
Molekylformel C11H13NO2
Kvantitet 25 mg
Synonym Benzyldiacetamide, N,N-Diacetylbenzylamine, N-Acetyl-N-(phenylmethyl)acetamide
InChI formel InChI=1S/C11H13NO2/c1-9(13)12(10(2)14)8-11-6-4-3-5-7-11/h3-7H,8H2,1-2H3
LEDER CC(=O)N(Cc1ccccc1)C(=O)C
IUPAC-namn N-acetyl-N-benzylacetamide
Molekylvikt (g/mol) 191.23
Formel vikt 191.0946