Learn More
N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog Number p-7077257
Click to view available options
Quantity:
1 g
5 g
25 g
29601-98-7
C7H9NO·ClH
159.62
YSNXOQGDHGUKCZ-UHFFFAOYSA-N
n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
11332622
N-benzylhydroxylamine;hydrochloride
C1=CC=C(C=C1)CNO.Cl
29601-98-7
C7H9NO·ClH
159.62
YSNXOQGDHGUKCZ-UHFFFAOYSA-N
n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
11332622
N-benzylhydroxylamine;hydrochloride
C1=CC=C(C=C1)CNO.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass Bottle
Linear Formula C6H5CH2NHOH·HCl
Quantity 1 g
Formula Weight 159.62
Percent Purity 98%
Chemical Name or Material N-Benzylhydroxylamine hydrochloride