Läs mer
N-Benzylidene-DL-phenylglycine Ethyl Ester, TRC
Produktkod. 30341656
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30341656 25 mg
1 options
Produktkod. 30341656 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB27955025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzylidene-DL-phenylglycine Ethyl Ester
CAS 77290-52-9
Molekylformel C17H17NO2
Kvantitet 25 mg
Synonym alpha-[(Phenylmethylene)amino]benzeneacetic Acid Ethyl Ester
InChI formel InChI=1S/C17H17NO2/c1-2-20-17(19)16(15-11-7-4-8-12-15)18-13-14-9-5-3-6-10-14/h3-13,16H,2H2,1H3
LEDER CCOC(=O)C(N=Cc1ccccc1)c2ccccc2
IUPAC-namn ethyl 2-(benzylideneamino)-2-phenylacetate
Molekylvikt (g/mol) 267.32
Formel vikt 267.1259