Läs mer
N-Benzylidene-L-alanine Sodium Salt, TRC
Produktkod. 30331799
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30331799 250 mg
1 options
Produktkod. 30331799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB635785250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzylidene-L-alanine Sodium Salt
CAS 113806-26-1
Molekylformel C10H10NNaO2
Kvantitet 250 mg
Synonym N-Phenylmethylene)-L-alanine Sodium Salt
InChI formel InChI=1S/C10H11NO2.Na/c1-8(10(12)13)11-7-9-5-3-2-4-6-9;/h2-8H,1H3,(H,12,13);/q;+1/p-1/t8-;/m0./s1
LEDER [Na+].C[C@H](N=Cc1ccccc1)C(=O)[O-]
IUPAC-namn sodium;(2S)-2-(benzylideneamino)propanoate
Molekylvikt (g/mol) 199.18
Formel vikt 199.0609