missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7054710
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
17400-34-9
C11H17ClN2O2
244.72
MFCD00270153
XKMBTMXQMDLSRB-UHFFFAOYSA-N
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
13196227
benzyl N-(3-aminopropyl)carbamate;hydrochloride
[H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
17400-34-9
C11H17ClN2O2
244.72
MFCD00270153
XKMBTMXQMDLSRB-UHFFFAOYSA-N
n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
13196227
benzyl N-(3-aminopropyl)carbamate;hydrochloride
[H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 185°C to 189°C
Color White
Quantity 1 g
Beilstein 4725495
Sensitivity Hygroscopic
Formula Weight 244.72
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'