Läs mer
N-(Benzyloxycarbonyl)aminodiacetic Acid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(Benzyloxycarbonyl)aminodiacetic Acid |
| CAS | 17335-88-5 |
| Molekylformel | C12H13NO6 |
| Kvantitet | 10 g |
| Synonym | N-(Carboxymethyl)-N-[(phenylmethoxy)carbonyl]-glycine, (Carboxyimino)di-acetic Acid N-Benzyl Ester, N-(Carboxymethyl)-N-[(phenylmethoxy)carbonyl]glycine |
| InChI formel | InChI=1S/C12H13NO6/c14-10(15)6-13(7-11(16)17)12(18)19-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,14,15)(H,16,17) |
| LEDER | OC(=O)CN(CC(=O)O)C(=O)OCc1ccccc1 |
| IUPAC-namn | 2-[carboxymethyl(phenylmethoxycarbonyl)amino]acetic acid |
| Molekylvikt (g/mol) | 267.23 |
| Formel vikt | 267.0743 |