Learn More
N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
Catalog Number 15448276
Click to view available options
Quantity:
250 mg
1 g
5 g
51644-83-8
C17H23NO6
337.372
MFCD00077018
GLMODRZPPBZPPB-CYBMUJFWSA-N
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
6992708
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
51644-83-8
C17H23NO6
337.372
MFCD00077018
GLMODRZPPBZPPB-CYBMUJFWSA-N
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
6992708
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 337.37
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester