Läs mer
N-Benzyloxycarbonyl-D-serine, TRC
Produktkod. 30332700
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30332700 250 mg
1 options
Produktkod. 30332700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB286730250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyloxycarbonyl-D-serine
CAS 6081-61-4
Molekylformel C11H13NO5
Kvantitet 250 mg
Synonym N-[(Phenylmethoxy)carbonyl]-D-serine, D-N-carboxy-Serine N-Benzyl ester, (Carbobenzyloxy)-D-serine, N-(Benzyloxycarbonyl)-D-serine, N-Carbobenzoxy-D-serine, N-Carbobenzyloxy-D-serine
InChI formel InChI=1S/C11H13NO5/c13-6-9(10(14)15)12-11(16)17-7-8-4-2-1-3-5-8/h1-5,9,13H,6-7H2,(H,12,16)(H,14,15)/t9-/m1/s1
LEDER OC[C@@H](NC(=O)OCc1ccccc1)C(=O)O
IUPAC-namn (2R)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
Molekylvikt (g/mol) 239.22
Formel vikt 239.0794