Läs mer
N-Benzyloxycarbonyl-Gamma-aminobutyric Acid, TRC
Produktkod. 30339317
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30339317 500 mg
1 options
Produktkod. 30339317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB286075500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyloxycarbonyl-gamma-aminobutyric Acid
CAS 5105-78-2
Molekylformel C12H15NO4
Kvantitet 500 mg
Synonym 4-(Benzyloxycarbonylamino)butanoic Acid, 4-(Benzyloxycarbonylamino)butyric Acid, 4-[(N-Carbobenzoxy)amino]butyric Acid, Carbobenzoxy-4-aminobutyric Acid, N-Benzyloxycarbonyl-gamma-aminobutyric Acid, N-Carbobenzyloxy-gamma-aminobutyric Acid, NSC 120007
InChI formel InChI=1S/C12H15NO4/c14-11(15)7-4-8-13-12(16)17-9-10-5-2-1-3-6-10/h1-3,5-6H,4,7-9H2,(H,13,16)(H,14,15)
LEDER OC(=O)CCCNC(=O)OCc1ccccc1
IUPAC-namn 4-(phenylmethoxycarbonylamino)butanoic acid
Molekylvikt (g/mol) 237.25
Formel vikt 237.1001