Läs mer
N-(Benzyloxycarbonyl)-L-Beta-chloroalanine Methyl Ester, TRC
Produktkod. 30333203
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30333203 100 mg
1 options
Produktkod. 30333203 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB285901100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Benzyloxycarbonyl)-L-beta-chloroalanine Methyl Ester
CAS 62107-38-4
Molekylformel C12 H14 Cl N O4
Kvantitet 100 mg
Synonym N-(Benzyloxycarbonyl)-L-Ss-Chloroalanine Methyl Ester
InChI formel InChI=1S/C12H14ClNO4/c1-17-11(15)10(7-13)14-12(16)18-8-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3,(H,14,16)/t10-/m0/s1
LEDER COC(=O)[C@H](CCl)NC(=O)OCc1ccccc1
IUPAC-namn methyl (2R)-3-chloro-2-(phenylmethoxycarbonylamino)propanoate
Molekylvikt (g/mol) 271.7
Formel vikt 271.0611