Läs mer
N-(Benzyloxycarbonyl)-L-glutamic Acid alpha-tert-Butyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(Benzyloxycarbonyl)-L-glutamic Acid alpha-tert-Butyl Ester |
| CAS | 5891-45-2 |
| Molekylformel | C17 H23 N O6 |
| Kvantitet | 1 g |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-glutamic Acid 1-(1,1-Dimethylethyl) Ester, N-Benzyl-N-carboxyglutamic acid 1-tert-Butyl Ester, NSC 169169, Z-Glu-OtBu, α-tert-Butyl N-benzyloxycarbonyl-L-glutamate |
| InChI formel | InChI=1S/C17H23NO6/c1-17(2,3)24-15(21)13(9-10-14(19)20)18-16(22)23-11-12-7-5-4-6-8-12/h4-8,13H,9-11H2,1-3H3,(H,18,22)(H,19,20)/t13-/m0/s1 |
| LEDER | CC(C)(C)OC(=O)[C@H](CCC(=O)O)NC(=O)OCc1ccccc1 |
| IUPAC-namn | (4S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid |
| Molekylvikt (g/mol) | 337.368 |
| Formel vikt | 337.153 |