Läs mer
N-Benzyloxycarbonyl L-Threonine Amide O-Methanesulfonate, TRC
Produktkod. 30344105
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30344105 250 mg
1 options
Produktkod. 30344105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB285924250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzyloxycarbonyl Threonine Amide O-Mesylate
CAS 80082-51-5
Molekylformel C13 H18 N2 O6 S
Kvantitet 250 mg
InChI formel InChI=1S/C13H18N2O6S/c1-9(21-22(2,18)19)11(12(14)16)15-13(17)20-8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3,(H2,14,16)(H,15,17)/t9-,11+/m1/s1
LEDER C[C@@H](OS(=O)(=O)C)[C@H](NC(=O)OCc1ccccc1)C(=O)N
IUPAC-namn [(2R,3S)-4-amino-4-oxo-3-(phenylmethoxycarbonylamino)butan-2-yl] methanesulfonate
Molekylvikt (g/mol) 330.36
Formel vikt 330.0886