missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7054831
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3496-11-5
MFCD00053547
This item is not returnable. View return policy
3496-11-5
MFCD00053547

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

Solubility
Soluble in 1mmol in 2mL DCM.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 114°C to 118°C
Color White
Molecular Formula C17H27ClN8O5
Beilstein 1555287
Synonym blasticidin s hcl
Solubility Information Soluble in 1mmol in 2mL DCM.
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
SMILES CN(CCC(CC(=O)NC1C=CC(OC1C(=O)O)N2C=CC(=NC2=O)N)N)C(=N)N.Cl
IUPAC Name (2R,3S,6S)-3-[[(3R)-3-amino-5-[carbamimidoyl(methyl)amino]pentanoyl]amino]-6-(4-amino-2-oxopyrimidin-1-yl)-3,6-dihydro-2H-pyran-2-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 458.90
PubChem CID 124080960
Formula Weight 348.35
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-L-valine N-succinimidyl ester
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'