Läs mer
N-Benzyloxycarbonyl Lisinopril Cyclohexyl Analogue Ethyl Methyl Diester, TRC
Produktkod. 30337888
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30337888 1 mg
1 options
Produktkod. 30337888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2857101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyloxycarbonyl Lisinopril Cyclohexyl Analogue Ethyl Methyl Diester
CAS 1356930-73-8
Molekylformel C32H49N3O7
Kvantitet 1 mg
Synonym (S)-1-[N2-(1-Carboxy-3-cyclohexylpropyl)-N-benzyloxycarbonyl-L-lysyl]-L-proline Ethyl Methyl Diester
InChI formel InChI=1S/C32H49N3O7/c1-3-41-30(37)27(20-19-24-13-6-4-7-14-24)34-26(29(36)35-22-12-18-28(35)31(38)40-2)17-10-11-21-33-32(39)42-23-25-15-8-5-9-16-25/h5,8-9,15-16,24,26-28,34H,3-4,6-7,10-14,17-23H2,1-2H3,(H,33,39)/t26-,27-,28-/m0/s1
LEDER CCOC(=O)[C@H](CCC1CCCCC1)N[C@@H](CCCCNC(=O)OCc2ccccc2)C(=O)N3CCC[C@H]3C(=O)OC
IUPAC-namn methyl (2S)-1-[(2S)-2-[[(2S)-4-cyclohexyl-1-ethoxy-1-oxobutan-2-yl]amino]-6-(phenylmethoxycarbonylamino)hexanoyl]pyrrolidine-2-carboxylate
Molekylvikt (g/mol) 587.75
Formel vikt 587.3571