Läs mer
N-Benzyloxycarbonylglycine Thioamide, TRC
Produktkod. 30346971
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30346971 50 mg
1 options
Produktkod. 30346971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB70018350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Benzyloxycarbonylglycine Thioamide
CAS 49548-40-5
Molekylformel C10H12N2O2S
Kvantitet 50 mg
Synonym Benzyl (2-amino-2-thioxoethyl) Carbamate, Phenylmethyl (2-Amino-2-thioxoethyl)carbamate, Phenylmethyl N-(2-Amino-2-thioxoethyl)carbamate
InChI formel InChI=1S/C10H12N2O2S/c11-9(15)6-12-10(13)14-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H2,11,15)(H,12,13)
LEDER NC(=S)CNC(=O)OCc1ccccc1
IUPAC-namn benzyl N-(2-amino-2-sulfanylideneethyl)carbamate
Molekylvikt (g/mol) 224.28
Formel vikt 224.0619