Learn More
N-Benzylprop-2-en-1-amine, 97%, BLD Pharm™
Product Code. 30988303
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30988303 25 g 25g
30974347 5 g -
30978208 1 g 1g
30972951 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30988303 Supplier BLD Pharm Supplier No. BD14314325g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 4383-22-6
Molecular Formula C10H13N
MDL Number MFCD00463446
Quantity 25 g
SMILES C=CCNCC1=CC=CC=C1
Molecular Weight (g/mol) 147.22
Percent Purity 97%
Chemical Name or Material N-Benzylprop-2-en-1-amine