Läs mer
N-Biotinyl p-Aminophenyl Arsenic Acid, TRC
Produktkod. 30345802
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30345802 5 mg
1 options
Produktkod. 30345802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3949705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Biotinyl p-Aminophenyl Arsenic Acid
CAS 212391-23-6
Anteckningar om renhetsgrad HPLC
Molekylformel C16H22AsN3O4S
Kvantitet 5 mg
Synonym As-[4-[[5-[(3aS,4, S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]phenyl]arsenic Acid
LEDER O=C(NC1=CC=C([As](O)O)C=C1)CCCC[C@H]2C3C(NC(N3)=O)CS2
Molekylvikt (g/mol) 427.35
Formel vikt 427.0547