Läs mer
N-Biotinylaminoethyl Methanethiosulfonate, TRC
Produktkod. 30348360
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30348360 10 mg
30331529 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30348360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB39475010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Biotinylaminoethyl Methanethiosulfonate
CAS 162758-04-5
Molekylformel C13 H23 N3 O4 S3
Kvantitet 10 mg
InChI formel InChI=1S/C13H23N3O4S3/c1-23(19,20)22-7-6-14-11(17)5-3-2-4-10-12-9(8-21-10)15-13(18)16-12/h9-10,12H,2-8H2,1H3,(H,14,17)(H2,15,16,18)/t9-,10-,12-/m0/s1
LEDER CS(=O)(=O)SCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC-namn 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(2-methylsulfonylsulfanylethyl)pentanamide
Molekylvikt (g/mol) 381.53
Formel vikt 381.0851