Läs mer
N-Biotinylcaproylaminocaproylaminocaproyl N-Hydroxysuccinimide, TRC
Produktkod. 30345920
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30345920 5 mg
1 options
Produktkod. 30345920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3957185MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Biotinylcaproylaminocaproylaminocaproyl N-Hydroxysuccinimide
CAS 1322625-23-9
Molekylformel C32 H52 N6 O8 S
Kvantitet 5 mg
InChI formel InChI=1S/C32H52N6O8S/c39-25(13-5-2-10-20-35-27(41)15-8-7-12-24-31-23(22-47-24)36-32(45)37-31)33-19-9-1-4-14-26(40)34-21-11-3-6-16-30(44)46-38-28(42)17-18-29(38)43/h23-24,31H,1-22H2,(H,33,39)(H,34,40)(H,35,41)(H2,36,37,45)/t23-,24-,31-/m0/s1
LEDER O=C(CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NCCCCCC(=O)NCCCCCC(=O)ON3C(=O)CCC3=O
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 6-[6-[6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoylamino]hexanoylamino]hexanoate
Molekylvikt (g/mol) 680.856
Formel vikt 680.357