Läs mer
N-Biotinylcaproylaminoethyl Methanethiosulfonate, TRC
Produktkod. 30343509
Change view
Kvantitet:
2 mg
Produktkod. Kvantitet
30343509 2 mg
1 options
Produktkod. 30343509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3957502MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Biotinylcaproylaminoethyl Methanethiosulfonate
CAS 353754-95-7
Molekylformel C19 H34 N4 O5 S3
Kvantitet 2 mg
InChI formel InChI=1S/C19H34N4O5S3/c1-31(27,28)30-12-11-21-17(25)8-3-2-6-10-20-16(24)9-5-4-7-15-18-14(13-29-15)22-19(26)23-18/h14-15,18H,2-13H2,1H3,(H,20,24)(H,21,25)(H2,22,23,26)/t14-,15-,18-/m0/s1
LEDER CS(=O)(=O)SCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC-namn 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-N-(2-methylsulfonylsulfanylethyl)hexanamide
Molekylvikt (g/mol) 494.69
Formel vikt 494.1691