Läs mer
N-Boc 1-O-t-Butyl 5-O-Methoxy L-Glutamic Acid, TRC
Produktkod. 30332696
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30332696 25 mg
1 options
Produktkod. 30332696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB64400525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Boc 1-O-t-Butyl 5-O-Methoxy L-Glutamic Acid
CAS 24277-38-1
Molekylformel C15H27NO6
Kvantitet 25 mg
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-L-glutamic Acid 1-(1,1-Dimethylethyl) 5-Methyl Ester, N-Carboxy-L-glutamic Acid N,1-Di-t-butyl Methyl Ester
InChI formel InChI=1S/C15H27NO6/c1-14(2,3)21-12(18)10(8-9-11(17)20-7)16-13(19)22-15(4,5)6/h10H,8-9H2,1-7H3,(H,16,19)/t10-/m0/s1
LEDER COC(=O)CC[C@H](NC(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC-namn 1-O-tert-butyl 5-O-methyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate
Molekylvikt (g/mol) 317.38
Formel vikt 317.1838