Läs mer
N-Boc-2-(2-naphthyl)-D-alanine, 99%, Thermo Scientific™
Produktkod. 15488427
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15488427 5 g 5g
15478427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15488427 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H51968.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76985-10-9
Molekylformel C18H21NO4
Molekylvikt (g/mol) 315.37
MDL-nummer MFCD00076900
InChI-nyckel URKWHOVNPHQQTM-OAHLLOKOSA-N
Synonym boc-d-2-nal-oh, boc-3-2-naphthyl-d-alanine, n-tert-butoxycarbonyl-3-2-naphthyl-d-alanine, boc-2-d-nal-oh, r-n-boc-2-naphthylalanine, boc-d-3-2-naphthyl-alanine, n-boc-3-2-naphthyl-d-alanine, n-boc-2-2-naphthyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
PubChem CID 2734481
IUPAC-namn (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
LEDER CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O
Specifikationer
Kemiskt namn eller material N-Boc-2-(2-naphthyl)-D-alanine
Smältpunkt 94°C to 96°C
Beilstein 4200660
Kvantitet 5 g
Formel vikt 315.37
Optisk rotation −45° (c=1 in Ethanol)
Procent renhet 99%