missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-2,4,5-trifluoro-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H62878.06

Additional Details : CAS Number : 486460-09-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449328

  • 7626.42 SEK / 5g


Estimated Shipment date
from Supplier 03-09-2018
Add to basket

Specifications

Chemical Identifiers

486460-09-7
319.28
FTWHJNNFVISGNQ-LLVKDONJSA-N
7146200
C14H16F3NO4
MFCD06659118
(R)-2-(Boc-amino)-3-(2,4,5-trifluorophenyl)propionic acid; Boc-D-2,4,5-trifluorophe

Specifications

N-Boc-2,4,5-trifluoro-D-phenylalanine
98%
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(2,4,5-trifluorophenyl)propanoic acid
7146200
Solid
486460-09-7
C14H16F3NO4
(R)-2-(Boc-amino)-3-(2,4,5-trifluorophenyl)propionic acid; Boc-D-2,4,5-trifluorophe
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
319.28
319.28
5g
Videos
SDS