Learn More
Alfa Aesar™ N-Boc-2-bromo-L-phenylalanine, 95%
Brand:  Alfa Aesar™ H62081.03
Additional Details : CAS Number : 261165-02-0 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15458668
  • 1504.07 SEK / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
261165-02-0
344.205
XDJSTMCSOXSTGZ-NSHDSACASA-N
2734475
C14H18BrNO4
MFCD01317709
(S)-N-Boc-2-bromophenylalanine; Boc-Phe(2-Br)-OH
Specifications
N-Boc-2-bromo-L-phenylalanine
261165-02-0
95%
MFCD01317709
XDJSTMCSOXSTGZ-NSHDSACASA-N
(2S)-3-(2-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2734475
1g
344.2
Solid
C14H18BrNO4
(S)-N-Boc-2-bromophenylalanine; Boc-Phe(2-Br)-OH
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Br)C(=O)O
344.205
139° to 140°C