Learn More
Alfa Aesar™ N-Boc-3-bromo-DL-phenylalanine, 97%
Brand:  Alfa Aesar™ H62935.06
Additional Details : CAS Number : 82278-95-3 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15469378
  • 4714.69 SEK / 5g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
N-Boc-3-bromo-DL-phenylalanine
344.2
3-Bromo-N-[(1,1-dimethylethoxy)carbonyl]-DL-Phenylalanine
82278-95-3
MFCD06656171
Specifications
N-Boc-3-bromo-DL-phenylalanine
Solid
344.2
3-Bromo-N-[(1,1-dimethylethoxy)carbonyl]-DL-Phenylalanine
82278-95-3
97%
MFCD06656171
5g