Läs mer
N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde, TRC
Produktkod. 30331360
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30331360 5 mg
1 options
Produktkod. 30331360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6615755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde
CAS 135865-78-0
Molekylformel C14 H19 N O3
Kvantitet 5 mg
Synonym Carbamic acid, N-[(1S)-3-oxo-1-phenylpropyl]-, 1,1-dimethylethyl ester, Carbamic acid, (3-oxo-1-phenylpropyl)-, 1,1-dimethylethyl ester, (S)-, Carbamic acid, [(1S)-3-oxo-1-phenylpropyl]-, 1,1-dimethylethyl ester (9CI), (S)-(3-Oxo-1-phenylpropyl)carbamic acid tert-butyl ester, (S)-3-Phenyl-3-Boc-aminopropionaldehyde, (S)-tert-Butyl (3-oxo-1-phenylpropyl)carbamate, tert-Butyl (1S)-3-oxo-1-phenylpropylcarbamate, N-Boc-(3S)-3-phenyl-3-aminopropionaldehyde, Boc-(S)-3-Amino-3-phenylpropanal, N-Boc-(3S)-3-Amino-3-phenylpropanal
InChI formel InChI=1S/C14H19NO3/c1-14(2,3)18-13(17)15-12(9-10-16)11-7-5-4-6-8-11/h4-8,10,12H,9H2,1-3H3,(H,15,17)/t12-/m0/s1
LEDER CC(C)(C)OC(=O)N[C@@H](CC=O)c1ccccc1
IUPAC-namn tert-butyl N-[(1S)-3-oxo-1-phenylpropyl]carbamate
Molekylvikt (g/mol) 249.31
Formel vikt 249.1365