Learn More
N-BOC-4-methylene-L-proline, 97%, Thermo Scientific Chemicals
Catalog Number p-7077327
Click to view available options
Quantity:
1 g
5 g
84348-38-9
C11H17NO4
227.26
MFCD01861787
ULLGRIBXGPATMA-UHFFFAOYNA-N
n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline
21727417
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O
84348-38-9
C11H17NO4
227.26
MFCD01861787
ULLGRIBXGPATMA-UHFFFAOYNA-N
n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline
21727417
(2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 110.0°C to 115.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-BOC-4-methylene-L-proline
compliance-icons
Product Identifier
  • N-BOC-4-methylene-L-proline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.