missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-norvaline dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog Number 15498368
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57521-85-4
C10H19NO4
217.265
MFCD11041002
INWOAUUPYIXDHN-SSDOTTSWSA-N
boc-d-norvaline, boc-d-nva-oh, r-2-tert-butoxycarbonyl amino pentanoic acid, 2r-2-tert-butoxycarbonyl amino pentanoic acid, boc-d-2-aminovaleric acid, n-t-butoxycarbonyl-d-norvaline, n-tert-butoxycarbonyl-d-norvaline, boc-d-nva-oh tlc, r-2-tert-butoxycarbonylamino-pentanoic acid, d-norvaline, n-1,1-dimethylethoxy carbonyl
6995000
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
57521-85-4
C10H19NO4
217.265
MFCD11041002
INWOAUUPYIXDHN-SSDOTTSWSA-N
boc-d-norvaline, boc-d-nva-oh, r-2-tert-butoxycarbonyl amino pentanoic acid, 2r-2-tert-butoxycarbonyl amino pentanoic acid, boc-d-2-aminovaleric acid, n-t-butoxycarbonyl-d-norvaline, n-tert-butoxycarbonyl-d-norvaline, boc-d-nva-oh tlc, r-2-tert-butoxycarbonylamino-pentanoic acid, d-norvaline, n-1,1-dimethylethoxy carbonyl
6995000
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 398.58
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Boc-D-norvaline dicyclohexylammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'