Läs mer
N-Boc-D-phenylalanine methyl ester, 98%
Produktkod. p-7054998
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15460179 1 g 1g
15470179 5 g 5g
15480179 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 15460179 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H63466.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 77119-84-7 | C15H21NO4 | 279.34 g/mol
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77119-84-7
Molekylformel C15H21NO4
Molekylvikt (g/mol) 279.34
MDL-nummer MFCD00672509
InChI-nyckel SDSWSVBXRBXPRL-UHFFFAOYNA-N
Synonym boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
PubChem CID 637614
LEDER COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C
Specifikationer
Kemiskt namn eller material N-Boc-D-phenylalanine methyl ester
Löslighetsinformation Slightly soluble in water.
IUPAC-namn methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
Kvantitet 1 g
Formel vikt 279.34
Procent renhet 98%
Fysisk form Solid