missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-phenylalanine methyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 15460179
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
77119-84-7
C15H21NO4
279.34
MFCD00672509
SDSWSVBXRBXPRL-UHFFFAOYNA-N
boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
637614
COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
77119-84-7
C15H21NO4
279.34
MFCD00672509
SDSWSVBXRBXPRL-UHFFFAOYNA-N
boc-d-phenylalanine methyl ester, boc-d-phe-ome, r-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-dphe-ome, n-tert-butoxycarbonyl-d-phenylalanine methyl ester, d-phenylalanine,n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoate
637614
COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C

N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-phenylalanine methyl ester employed as a primary and secondary intermediate chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Solubility Information Slightly soluble in water.
IUPAC Name methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
Formula Weight 279.34
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Boc-D-phenylalanine methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'