missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-D-phenylalanine N-succinimidyl ester, 97%

Brand:  Alfa Aesar™ H62763.14

Additional Details : CAS Number : 3674-18-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15449248

  • 6674.87 SEK / 25g


Estimated Shipment date
from Supplier 17-12-2018
Add to basket

Specifications

Chemical Identifiers

3674-18-8
362.382
NHUCANAMPJGMQL-CYBMUJFWSA-N
11164339
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
C18H22N2O6
MFCD00069688
boc-d-phe-osu, boc-d-phenylalanine n-hydroxysuccinimide ester, r-2,5-ddioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-d-phenylalanine n-hydroxy succinimide ester, n-tert-butoxycarbonyl-d-phenylalanine succinimidyl ester, n-alpha-t-butyloxycarbonyl-d-phenylalanine succinimidyl ester, boc-d-phe-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2r-2-tert-butoxy carbonylamino-3-phenylpropanoate, 2,5-dioxopyrrolidin-1-yl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate
(2,5-dioxopyrrolidin-1-yl) (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate

Specifications

N-Boc-D-phenylalanine N-succinimidyl ester
C18H22N2O6
1555676
NHUCANAMPJGMQL-CYBMUJFWSA-N
(2,5-dioxopyrrolidin-1-yl) (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
11164339
Powder
White
25g
3674-18-8
MFCD00069688
boc-d-phe-osu, boc-d-phenylalanine n-hydroxysuccinimide ester, r-2,5-ddioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, boc-d-phenylalanine n-hydroxy succinimide ester, n-tert-butoxycarbonyl-d-phenylalanine succinimidyl ester, n-alpha-t-butyloxycarbonyl-d-phenylalanine succinimidyl ester, boc-d-phe-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2r-2-tert-butoxy carbonylamino-3-phenylpropanoate, 2,5-dioxopyrrolidin-1-yl 2r-2-tert-butoxycarbonyl amino-3-phenylpropanoate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
362.382
362.38
97%
149°C to 153°C
Moisture Sensitive
Videos
SDS