Läs mer
N-Boc-erythro-sphingosine, TRC
Produktkod. 30179066
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179066 250 mg
30179241 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB667750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 116467-63-1
Molekylformel C23 H45 N O4
Molekylvikt (g/mol) 399.61
IUPAC-namn tert-butyl N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]carbamate
LEDER CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(=O)OC(C)(C)C
Specifikationer
Kemiskt namn eller material N-Boc-erythro-sphingosine
InChI formel InChI=1S/C23H45NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-21(26)20(19-25)24-22(27)28-23(2,3)4/h17-18,20-21,25-26H,5-16,19H2,1-4H3,(H,24,27)/b18-17+/t20-,21+/m0/s1
Kvantitet 250 mg
Formel vikt 399.3349