Learn More
N-Boc-glycine methyl ester, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Description
It is used as a pharmaceutical, chemical, paint and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical, chemical, paint and dye intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is used as a pharmaceutical, chemical, paint and dye intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Density | 1.079 |
Boiling Point | 190°C |
Flash Point | 109°C (228°F) |
Linear Formula | (CH3)3COCONHCH2CO2CH3 |
Refractive Index | 1.437 |
Quantity | 5 mL |
Formula Weight | 189.21 |
Percent Purity | 97% |
Chemical Name or Material | N-Boc-glycine methyl ester |