Läs mer
N-Boc-L-alanine N-succinimidyl ester, 98%, Thermo Scientific™
Produktkod. 15415318
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15415318 5 g 5g
15405318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15415318 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H59104.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3392-05-0
Molekylformel C12H18N2O6
Molekylvikt (g/mol) 286.284
MDL-nummer MFCD00065974
InChI-nyckel COMUWNFVTWKSDT-ZETCQYMHSA-N
Synonym boc-ala-osu, succinimido s-2-tert-butoxycarbonyl amino propionate, boc-l-alanine n-succinimidyl ester, boc-l-alanine n-hydroxysuccinimide ester, s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino propanoate, boc-l-ala-osu, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino propanoate, boc-l-alanine n-hydroxysuccinimde ester, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-1-methyl-2-oxoethylcarbamate #, n-boc-l-alanine n-succinimidyl ester
PubChem CID 11011585
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
LEDER CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
Specifikationer
Kemiskt namn eller material N-Boc-L-alanine N-succinimidyl ester
Smältpunkt 161°C to 163°C
Kvantitet 5 g
Beilstein 3560476
Känslighet Moisture sensitive
Optisk rotation −52° (c=2.5 in Dioxane)
Formel vikt 286.28
Procent renhet 98%