missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-aspartic acid 1-tert-butyl ester, 95%

Brand:  Alfa Aesar™ H62425.03

Additional Details : CAS Number : 34582-32-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448928

  • 443.51 SEK / 1g


Estimated Shipment date
from Supplier 14-11-2018
Add to basket

Specifications

Chemical Identifiers

34582-32-6
289.328
RAUQRYTYJIYLTF-QMMMGPOBSA-N
7010517
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
C13H23NO6
MFCD00038272
boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu
(3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid

Specifications

N-Boc-L-aspartic acid 1-tert-butyl ester
C13H23NO6
4191701
RAUQRYTYJIYLTF-QMMMGPOBSA-N
(3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
7010517
Powder
White
Odorless
34582-32-6
MFCD00038272
boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
289.328
289.32
95%
1g
Videos
SDS