Learn More
N-BOC-L-Aspartic acid, 99%, Thermo Scientific™
Catalog Number p-4511392
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
13726-67-5
C9H15NO6
233.22
KAJBMCZQVSQJDE-YFKPBYRVSA-N
boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
99718
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
13726-67-5
C9H15NO6
233.22
KAJBMCZQVSQJDE-YFKPBYRVSA-N
boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
99718
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O
Melting Point 116.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2CCH2CH[NHCO2C(CH3)3]CO2H
Quantity 1 g
Specific Rotation − 6.20
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 233.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-L-Aspartic acid