missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-benzyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7038014
Click to view available options
Quantity:
5 g
25 g
13574-13-5
C17H22NO6
336.37
MFCD00065569
AJDUMMXHVCMISJ-ZDUSSCGKSA-M
boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
83589
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O
13574-13-5
C17H22NO6
336.37
MFCD00065569
AJDUMMXHVCMISJ-ZDUSSCGKSA-M
boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
83589
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O

N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-glutamic acid 5-benzyl ester is used as a pharmaceutical intermediate.

Solubility
Soluble in N,N- Dimethyl formamide.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Melting Point 69°C to 71°C
Quantity 5 g
Beilstein 2226572
Solubility Information Soluble in N,N- Dimethyl formamide.
Optical Rotation −5.5° (c=1 in Acetic Acid)
Formula Weight 337.37
Percent Purity 98%
Chemical Name or Material N-Boc-L-glutamic acid 5-benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'