missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-leucine methyl ester, 97%

Brand:  Alfa Aesar™ H61551.14

Additional Details : CAS Number : 63096-02-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15498108

  • 3777.83 SEK / 25g


Estimated Shipment date
from Supplier 21-05-2019
Add to basket

Specifications

Chemical Identifiers

63096-02-6
245.319
QSEVMIMUBKMNOU-VIFPVBQESA-N
10879384
CC(C)CC(C(=O)OC)NC(=O)OC(C)(C)C
C12H23NO4
MFCD00076952
boc-leu-ome, s-methyl 2-tert-butoxycarbonyl amino-4-methylpentanoate, n-tert-butoxycarbonyl-l-leucine methyl ester, n-boc-l-leucine methyl ester, boc-l-leucine methyl ester, l-leucine,n-1,1-dimethylethoxy carbonyl-, methyl ester, n-boc-leucine methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-4-methylpentanoate, boc-l-leucyl methyl ester, bidd:gt0678
methyl (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate

Specifications

N-Boc-L-leucine methyl ester
C12H23NO4
boc-leu-ome, s-methyl 2-tert-butoxycarbonyl amino-4-methylpentanoate, n-tert-butoxycarbonyl-l-leucine methyl ester, n-boc-l-leucine methyl ester, boc-l-leucine methyl ester, l-leucine,n-1,1-dimethylethoxy carbonyl-, methyl ester, n-boc-leucine methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-4-methylpentanoate, boc-l-leucyl methyl ester, bidd:gt0678
1.4400
methyl (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
10879384
Liquid
0.991g/mL
25g
63096-02-6
MFCD00076952
QSEVMIMUBKMNOU-VIFPVBQESA-N
CC(C)CC(C(=O)OC)NC(=O)OC(C)(C)C
245.319
245.32
97%
205°C
Videos
SDS