Learn More
N-Boc-L-leucinol, 97%, Thermo Scientific Chemicals
Catalog Number p-7047508
Click to view available options
Quantity:
1 g
5 g
82010-31-9
C11H23NO3
217.31
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C
82010-31-9
C11H23NO3
217.31
MFCD00076950
LQTMEOSBXTVYRM-VIFPVBQESA-N
boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
7018766
tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 212°C to 214°C
Beilstein 4178460
Refractive Index 1.45
Formula Weight 217.31
Percent Purity 97%
Density 0.98
Quantity 1 g
Flash Point >110°C (230°F)
Optical Rotation −27.5° (c=2 in Methanol)
Chemical Name or Material N-Boc-L-leucinol