missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-methionine sulfoxide, 98%

Brand:  Alfa Aesar™ H63053.06

Additional Details : CAS Number : 34805-21-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449518

  • 526.43 SEK / 5g


Estimated Shipment date
from Supplier 16-11-2018
Add to basket

Specifications

Chemical Identifiers

34805-21-5
265.324
FVSDTYGQCVACMH-ISJKBYAMSA-N
7408090
CC(C)(C)OC(=O)NC(CCS(=O)C)C(=O)O
C10H19NO5S
MFCD00037172
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid

Specifications

N-Boc-L-methionine sulfoxide
C10H19NO5S
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
CC(C)(C)OC(=O)NC(CCS(=O)C)C(=O)O
265.324
265.33
98%
34805-21-5
MFCD00037172
FVSDTYGQCVACMH-ISJKBYAMSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
7408090
Solid
5g
Videos
SDS