missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-pipecolinic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Melting Point | 121°C to 125°C |
Quantity | 250 mg |
Beilstein | 3652038 |
Solubility Information | Insoluble in water. |
Optical Rotation | −46° (c=1 in Methanol) |
Formula Weight | 229.28 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Boc-L-pipecolinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'