missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-serine hydrate, 98%

Brand:  Alfa Aesar™ H62458.MD

Additional Details : CAS Number : 204191-40-2 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15498968

  • 357.62 SEK / 250mg


Estimated Shipment date
from Supplier 15-07-2019
Add to basket

Specifications

Chemical Identifiers

204191-40-2
223.225
JLIWGTMHYWJUPM-JEDNCBNOSA-N
51358356
CC(C)(C)OC(=O)NC(CO)C(=O)O.O
C8H17NO6
MFCD00150501
s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid hydrate, boc-l-serine hydrate, boc-l-serine monohydrate, 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid hydrate, boc-ser-oh. h2o, pubchem12002, n-tert-butoxycarbonyl-l-serine-water 1/1, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoicacidhydrate
(2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid;hydrate

Specifications

N-Boc-L-serine hydrate
C8H17NO6
s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid hydrate, boc-l-serine hydrate, boc-l-serine monohydrate, 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid hydrate, boc-ser-oh. h2o, pubchem12002, n-tert-butoxycarbonyl-l-serine-water 1/1, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoicacidhydrate
CC(C)(C)OC(=O)NC(CO)C(=O)O.O
223.225
223.22
98%
204191-40-2
MFCD00150501
JLIWGTMHYWJUPM-JEDNCBNOSA-N
(2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid;hydrate
51358356
Solid
250mg
Videos
SDS