missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-serine hydrate, 98%

Brand:  Alfa Aesar™ H62458.03

Additional Details : CAS Number : 204191-40-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478968

  • 884.12 SEK / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-Boc-L-serine hydrate
223.22
Boc-Ser-OH.H2O; (S)-2-(Boc-amino)-3-hydroxypropionic acid hydrate
204191-40-2
MFCD00150501

Specifications

N-Boc-L-serine hydrate
Solid
223.22
Boc-Ser-OH.H2O; (S)-2-(Boc-amino)-3-hydroxypropionic acid hydrate
204191-40-2
98%
MFCD00150501
1g
Videos
SDS