Learn More
Alfa Aesar™ N-Boc-L-styrylalanine dicyclohexylammonium salt, 95%
Brand:  Alfa Aesar™ H62167.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15477106
  • 8147.06 SEK / 5g
Estimated Shipment date
from Supplier 06-03-2018
Add to basket
Specifications
Chemical Identifiers
N-Boc-L-styrylalanine dicyclohexylammonium salt
261165-04-2
472.66
MFCD01860895
Specifications
N-Boc-L-styrylalanine dicyclohexylammonium salt
Powder
472.66
White
261165-04-2
95%
MFCD01860895
5g